Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR graduated laborator1EA
Catalog No. 502909129
Encompass
Reusable bottles without caps or pouring rings for the storage of reagents chemicals SOLUTIONons tissue culture media etc. Made of Duran borosilicate glass Type 3.3 for maximum chemical and thermal shock resistance.Optional pouring rings prevent drips when pouring from these bottles.
Catalog No. 50-290-9129 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2322621EA
Reusable bottles without caps or pouring rings for the storage of reagents chemicals SOLUTIONons tissue culture media etc. Made of Duran borosilicate glass Type 3.3 for maximum chemical and thermal shock resistance.Optional pouring rings prevent drips when pouring from these bottles.