Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR laboratory bottles
Catalog No. 502901424
Encompass
Reusable bottles and caps for the storage of reagents chemicals SOLUTIONons tissue culture media etc. Made of Duran borosilicate glass Type 3.3 for maximum chemical and thermal shock resistance.
Catalog No. 50-290-1424 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z30518910EA
Reusable bottles and caps for the storage of reagents chemicals SOLUTIONons tissue culture media etc. Made of Duran borosilicate glass Type 3.3 for maximum chemical and thermal shock resistance.