Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR plastic coated labo
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z15390710EA
Encompass
Transparent plastic skin protects bottles from mechanical damage and leakage in case of breakage. Autoclavable to 135C but do not use water for cooling in autoclave.Please note Cap not included.
Catalog No. 50-290-9087