Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR square bottle clear
Catalog No. 502901010
Encompass
With easy-to-read scale and large labeling field for easy marking. Complete with blue screw cap and pouring ring for drip-free pouring and clean safe working. Service temperature limit of screw cap and pouring ring 140C.Ergonomic handling due to angular shape highly stable good stackabilityProvides a space saving of 44% in comparison with standard laboratory bottles
Catalog No. 50-290-1010 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z68110510EA
With easy-to-read scale and large labeling field for easy marking. Complete with blue screw cap and pouring ring for drip-free pouring and clean safe working. Service temperature limit of screw cap and pouring ring 140C.Ergonomic handling due to angular shape highly stable good stackabilityProvides a space saving of 44% in comparison with standard laboratory bottles