Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR super duty beaker c1EA
Catalog No. 502906031
Encompass
To avoid thermal stressing of the glass uniform and slow heating of super duty products is recommended.When working at very high temperatures or if rapid temperature changes are expected standard beakers and Erlenmeyer flasks should be used as these are characterized by excellent thermal shock resistance.However the mechanical sturdiness of standard products is limited compared with the super duty range.bullnbspIncreased mechanical sturdiness due to increased uniform wall thicknessbullnbspReinforced sturdy rimbullnbspType 1 glassbullnbspAutoclavable/sterilizable
Catalog No. 50-290-6031 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6809581EA
To avoid thermal stressing of the glass uniform and slow heating of super duty products is recommended.When working at very high temperatures or if rapid temperature changes are expected standard beakers and Erlenmeyer flasks should be used as these are characterized by excellent thermal shock resistance.However the mechanical sturdiness of standard products is limited compared with the super duty range.bullnbspIncreased mechanical sturdiness due to increased uniform wall thicknessbullnbspReinforced sturdy rimbullnbspType 1 glassbullnbspAutoclavable/sterilizable