Learn More
Research Products International Corp Duraporter Laboratory Transport Box, 15 x 7 3/4 x 6 1/4 inches High, Red with Red Handles
SDP
Catalog No. 501367702
Encompass
DuraPorter provides a secure and clean environment for carrying biological and clinical samples from one location to another. Ideal for use with a variety of laboratory items such as tubes, bottles, vials and instruments. DuraPorter transport boxes are constructed from high clarity polycarbonate that allows easy visibility of contents.
Transport boxes feature a positive latching hinged lid, leak tight silicone gasket, three integral clasps and a folding handle. Lid opens 180° for easy access to the interior compartment. Supplied with dividers to customize interior space.
Transport boxes can be autoclaved at 121°C for a maximum of 15 minutes.
Note: Handles should remain open during autoclaving process.
Catalog No. 50-136-7702 Supplier Research Products International Corp Supplier No. 246816
DuraPorter provides a secure and clean environment for carrying biological and clinical samples from one location to another. Ideal for use with a variety of laboratory items such as tubes, bottles, vials and instruments. DuraPorter transport boxes are constructed from high clarity polycarbonate that allows easy visibility of contents.
Transport boxes feature a positive latching hinged lid, leak tight silicone gasket, three integral clasps and a folding handle. Lid opens 180° for easy access to the interior compartment. Supplied with dividers to customize interior space.
Transport boxes can be autoclaved at 121°C for a maximum of 15 minutes.
Note: Handles should remain open during autoclaving process.