Learn More
Fisherbrand™ DuraPorter™ XL Tube Insert
Catalog No. 17183978 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
Blood Tubes (Large)
Blood Tubes (Small and Large)
Blood Tubes (Small)
Array:
10 x 10
12 x 14
5 x 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Array
17-183-978 Blood Tubes (Large) 10 x 10
17-183-977 Blood Tubes (Small) 12 x 14
17-183-976 Blood Tubes (Small and Large) 5 x 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-183-978
Add to Cart
Add to Cart
DuraPorter™ XL Tube Rack Insert designed to fit DuraPorter™ XL High-Capacity Sample Transport.
  • Designed to fit DuraPorter™ XL High-Capacity Sample Transport
  • Thermoformed PETG
  • Tube Rack Inserts can be used separately
  • Lid removes completely for easy cleaning
  • Insert deep enough to be filled with ice
Specifications
Autoclavable Not Autoclavable
Color Clear
Holds Blood Tubes (Large)
Material Thermoformed PETG
Positions 1
Shape Rectangular, Rounded Corners
Stackable Yes, When Empty
Sterility Non-sterile
Dimensions (L x W x H) 13.1 x 14.3 x 5.55 in.
Product Type Tube Holder
Shelves None
Array 10 x 10