Learn More
Sigma Aldrich Fine Chemicals Biosciences DuraPorterR specimen trans
Catalog No. 502900947
Encompass
The DuraPorter Specimen or Sample Transport Box is ideal for transporting biological and clinical samples as well as instruments and products that you want to keep clean and dry under testing environmental conditions. The clarity of the DuraPorter(R) enables the user to visibly check sample integrity so that it can be opened using the right precautions. Polycarbonate construction and avaiable in 3 colors for user/ ownership identification. Silicone water-tight seal and three lid clasps provide a secure closure. Carry handle and folds neatly into body space allowing the units to be stacked. Lid opens a full 180 allowing for total access to contents and making it easier to clean. Will accomodate 13 and 16mm tubes in a 72-place unwire rack or similar sized rack. Unit is autoclavable.
Catalog No. 50-290-0947 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS120052
The DuraPorter Specimen or Sample Transport Box is ideal for transporting biological and clinical samples as well as instruments and products that you want to keep clean and dry under testing environmental conditions. The clarity of the DuraPorter(R) enables the user to visibly check sample integrity so that it can be opened using the right precautions. Polycarbonate construction and avaiable in 3 colors for user/ ownership identification. Silicone water-tight seal and three lid clasps provide a secure closure. Carry handle and folds neatly into body space allowing the units to be stacked. Lid opens a full 180 allowing for total access to contents and making it easier to clean. Will accomodate 13 and 16mm tubes in a 72-place unwire rack or similar sized rack. Unit is autoclavable.