Learn More
Med Vet International 3M Duraprep Applicator, 26 mL
Supplier:  Med Vet International MMM8630H
Encompass_Preferred
Clinically proven and cost-effective, this patient skin prepping solution meets AORN and CDC guidelines for the reduction of surgical site infections. Benefits Is 40% less expensive than ChloraPrep Patient Preoperative Skin Preparation per 26 mL applicator{1}Designed to be applied in a single, painted coat -- 2 log reduction -- abdomen at 10 minutes -- 3 log reduction -- groin at 10 minutes -- Both sites maintain counts below baseline out to 6 hours. Covers more than twice the area than ChloraPrep. According to labeling, a 26 mL applicator of ChloraPrep will cover an area 13.2 inches by 13.2 inches, or 174 square inches. In comparison, a 26 mL applicator of 3M DuraPrep™ Surgical Prepping Solution will cover 15 inches by 30 inches or 450 square inches. Has substantial clinical documentation including data in simulated surgical conditions -- 5 human safety studies -- 9 in vitro studies -- 8 in vitro studies -- 8 clinical in-use studies.
Catalog No. 50-283-1179