Learn More
LGC GENOMICS LLC (Lucigen) DuraScribe T7 Transcription Kit, 25 RxnsProduct RNase-resistant transcripts for RNAi and SELEX applications.
Catalog No. NC2162042
Encompass
Produce fully RNase A-resistant RNA that is perfectly suited for RNA aptamer synthesis, as well as antisense RNA and RNA interference (RNAi) experiments.Protect transcribed RNA from RNAse degradationProduce high RNA yieldsSelect and optimize RNA aptamers using SELEX screeningContents: DuraScribe T7 Enzyme Mix, RNase-Free DuraScribe T7 10X Reaction Buffer, ATP, GTP, 2-F-dCTP, 2-F-dUTP, DNase I, DTT, Control Template DNA (linearised), Sterile Deionised Water.
This item is not returnable. View return policy
Catalog No. NC2162042 Supplier LGC GENOMICS LLC Supplier No. DS010925
Produce fully RNase A-resistant RNA that is perfectly suited for RNA aptamer synthesis, as well as antisense RNA and RNA interference (RNAi) experiments.Protect transcribed RNA from RNAse degradationProduce high RNA yieldsSelect and optimize RNA aptamers using SELEX screeningContents: DuraScribe T7 Enzyme Mix, RNase-Free DuraScribe T7 10X Reaction Buffer, ATP, GTP, 2-F-dCTP, 2-F-dUTP, DNase I, DTT, Control Template DNA (linearised), Sterile Deionised Water.