Learn More
LGC GENOMICS LLC (Lucigen) DuraScribe T7 Transcription Kit, 10 RxnsProduct RNase-resistant transcripts for RNAi and SELEX applications.
Supplier:  LGC GENOMICS LLC DS010910
Encompass
The DuraScribe T7 Transcription Kit produces 2'-Fluorine-modified RNA transcriptscalled DuraScript RNAthat are completely resistant to RNase A. Ideal for SELEX RNA-aptamer selection procedures.
Catalog No. NC0272791