The DuraScrim Jacket provides an economical yet durable solution with double-coated PVC on polyester, ensuring high tear strength. Flame-resistant and waterproof with sealed seams, it offers protection against contaminants while allowing freedom of movement.
- Double-coated PVC on polyester for durability and high tear strength
- Full-cut shoulder and 90° sleeve-to-body angle for mobility
- Storm fly front seals out wind, rain, and contaminants
- Non-conductive, non-corrosive hardware
- Enhanced abrasion resistance with thicker outer coating
- Mildew resistant
- ASTM F1671 passing result for viral penetration resistance
- Therm-O-Rad sealed seams for 100% waterproof and chemical splash protection
- Flame resistant per ASTM D6413, self-extinguishing
- Resists many acids, oils, alcohols, salts, and alkalis