Learn More
MSC DURASCRIM PVC3PC RAINSUIT
Catalog No. 5000794597
Encompass
The DuraScrim 3-Piece Suit offers durable, economical protection. Constructed from double-coated PVC on polyester with an open weave scrim, it provides high tear strength. This suit includes a jacket, detachable hood, and overalls.
  • Features a 90° sleeve-to-body angle and full-cut shoulder for freedom of movement
  • Storm fly front jacket for weather and contaminant protection
  • Self-material collar resists contamination
  • Thicker outside coating for better abrasion resistance
  • Fully waterproof construction
  • Therm-O-Rad sealed seams for chemical splash protection
  • Achieved passing result for ASTM F1671 resistance to viral penetration
  • Flame resistant per ASTM D6413, self-extinguishing with flameout and afterglow less than or equal to 2 seconds
Catalog No. 50-007-94597 Supplier MSC Supplier No. 71502892
The DuraScrim 3-Piece Suit offers durable, economical protection. Constructed from double-coated PVC on polyester with an open weave scrim, it provides high tear strength. This suit includes a jacket, detachable hood, and overalls.
  • Features a 90° sleeve-to-body angle and full-cut shoulder for freedom of movement
  • Storm fly front jacket for weather and contaminant protection
  • Self-material collar resists contamination
  • Thicker outside coating for better abrasion resistance
  • Fully waterproof construction
  • Therm-O-Rad sealed seams for chemical splash protection
  • Achieved passing result for ASTM F1671 resistance to viral penetration
  • Flame resistant per ASTM D6413, self-extinguishing with flameout and afterglow less than or equal to 2 seconds