Learn More
Research Products International Corp DuraSeal Laboratory Stretch Film, 4 Inch x 250 Feet
SDP
Catalog No. 50212425
Encompass
DuraSeal is a durable, heat and solvent resistant stretch film with substantial advantages over common laboratory stretch films. Superior heat resistance allows it to be used for covering containers in boiling water baths. DuraSeal is polyethylene based and free of any adhesives. It resists essentially all organic solvents and caustic agents. The film can be stretched 500-800% before tearing, minimizing the incidence of tearing during stretch sealing.
DuraSeal is easy to use without the interleaf paper to strip away. Just stretch the film over a container opening. The film contracts inwardly to provide a drum tight liquid proof stretch seal. The film clings to itself, especially after stretching. DuraSeal cuts easily with sharp scissors or razor blade.
Catalog No. 50-212-425 Supplier Research Products International Corp Supplier No. 164250
DuraSeal is a durable, heat and solvent resistant stretch film with substantial advantages over common laboratory stretch films. Superior heat resistance allows it to be used for covering containers in boiling water baths. DuraSeal is polyethylene based and free of any adhesives. It resists essentially all organic solvents and caustic agents. The film can be stretched 500-800% before tearing, minimizing the incidence of tearing during stretch sealing.
DuraSeal is easy to use without the interleaf paper to strip away. Just stretch the film over a container opening. The film contracts inwardly to provide a drum tight liquid proof stretch seal. The film clings to itself, especially after stretching. DuraSeal cuts easily with sharp scissors or razor blade.