Learn More
Electron Microscopy Sciences DuraSeal™ Stretch Film, 12" width 250 FT
SDP
Catalog No. 501927388
Encompass_Preferred
A durable, heat-resistant, solvent-proof laboratory stretch film. Microwave compatible, withstanding temperatures up to 100 degree C (boiling water). Resists esssentially all solvents and caustic agents. Resists tearing and can stretch 500-800% before breaking or tearing. Film can cling to itself following stretching and forms an airtight barrier.
Width: 12"
Catalog No. 50-192-7388 Supplier Electron Microscopy Sciences Supplier No. 7099414
A durable, heat-resistant, solvent-proof laboratory stretch film. Microwave compatible, withstanding temperatures up to 100 degree C (boiling water). Resists esssentially all solvents and caustic agents. Resists tearing and can stretch 500-800% before breaking or tearing. Film can cling to itself following stretching and forms an airtight barrier.
Width: 12"