Learn More
Electron Microscopy Sciences DuraSeal™ Cling Sealing Film 1" wide 150 FT
SDP
Catalog No. 5034540
Encompass_Preferred
Film can cling to itself and glass vessels. It is heat Resistant and softens at 100 degree C. It is strong and will stretch 300-400% before breaking or tearing. It is tight sealing and forms and air tight barrier when stretched across openings. Ideal for Test Tubes.
Width: 1"
Catalog No. 50-345-40 Supplier Electron Microscopy Sciences Supplier No. 7099407
Film can cling to itself and glass vessels. It is heat Resistant and softens at 100 degree C. It is strong and will stretch 300-400% before breaking or tearing. It is tight sealing and forms and air tight barrier when stretched across openings. Ideal for Test Tubes.
Width: 1"