Learn More
Electron Microscopy Sciences DuraSeal™ Stretch Film, 1" width 150 FT
SDP
Catalog No. 5025666
Encompass_Preferred
A durable, heat-resistant, solvent-proof laboratory stretch film. Microwave compatible, withstanding temperatures up to 100 degree C (boiling water). Resists esssentially all solvents and caustic agents. Resists tearing and can stretch 500-800% before breaking or tearing. Film can cling to itself following stretching and forms an airtight barrier.
Width: 1"
Catalog No. 50-256-66 Supplier Electron Microscopy Sciences Supplier No. 7099401
A durable, heat-resistant, solvent-proof laboratory stretch film. Microwave compatible, withstanding temperatures up to 100 degree C (boiling water). Resists esssentially all solvents and caustic agents. Resists tearing and can stretch 500-800% before breaking or tearing. Film can cling to itself following stretching and forms an airtight barrier.
Width: 1"