Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Guard Cartridge 10 x 2.1mm

Catalog No. 502398382 Shop All Phenomenex Products
Encompass

Durashell 3m C18(L) 150 DC930102-LS

Catalog No. 50-239-8382 Supplier Phenomenex Inc Supplier No. DC930102LS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Durashell 3m C18(L) 150 DC930102-LS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.