Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Guard Cartridge 10 x 4.6mm

Catalog No. 502397885 Shop All Phenomenex Products
Encompass

Durashell 5m C18 150 DC950105-L

Catalog No. 50-239-7885 Supplier Phenomenex Inc Supplier No. DC950105L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Durashell 5m C18 150 DC950105-L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.