Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 250 x 21.2mm

Catalog No. 502394152 Shop All Phenomenex Products
Encompass

Durashell 5m C18(L) 150 DC952520-L

Catalog No. 50-239-4152 Supplier Phenomenex Inc Supplier No. DC952520L
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Durashell 5m C18(L) 150 DC952520-L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.