Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 50 x 2.1mm

Catalog No. 502394058 Shop All Phenomenex Products
Encompass

Durashell 5m C18(L) 150 DC950502-L

Catalog No. 50-239-4058 Supplier Phenomenex Inc Supplier No. DC950502L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Durashell 5m C18(L) 150 DC950502-L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.