Learn More
DURATHERM EXTENDED LIFE FLUIDS Duratherm S, 5 gallon pail
Catalog No. NC9652388
Encompass
A silicone based fluid that is ideally suited for high oxidation or completely exposed systems. Duratherm S is virtually impervious to oxidation at temperatures under 204C (400F) and offers superior protection from fluid degradation. Duratherm S is best suited for applications not prone to contamination or leaking.
Catalog No. NC9652388 Supplier DURATHERM EXTENDED LIFE FLUIDS Supplier No. DURATHERM S5GAL/PAIL
A silicone based fluid that is ideally suited for high oxidation or completely exposed systems. Duratherm S is virtually impervious to oxidation at temperatures under 204C (400F) and offers superior protection from fluid degradation. Duratherm S is best suited for applications not prone to contamination or leaking.