Learn More
Research Products International Corp DuraTube Micro-Tubes, 2.0ml Capacity, Includes Caps, 200 per Case
SDP
Catalog No. 50998565
Encompass
DuraTube is made of a specially formulated polymer for use in rugged bead beating applications. This resilient tube is shatter resistant and prevents sample loss during vigorous processing. Combination screw cap with o-ring ensures a tight seal and is supplied attached to tube. Free standing tube has a capacity of 2.0ml with graduated markings. Supplied 200 tubes and caps per case, non-sterile.
Catalog No. 50-998-565 Supplier Research Products International Corp Supplier No. 163283
DuraTube is made of a specially formulated polymer for use in rugged bead beating applications. This resilient tube is shatter resistant and prevents sample loss during vigorous processing. Combination screw cap with o-ring ensures a tight seal and is supplied attached to tube. Free standing tube has a capacity of 2.0ml with graduated markings. Supplied 200 tubes and caps per case, non-sterile.