Learn More
Electron Microscopy Sciences Durcupan Single Component B
SDP
Catalog No. 50980386
Encompass_Preferred
Single Component B Durcupan, Water Soluble- a water soluble embedding medium for EM based on an aliphatic polyepoxide.
Catalog No. 50-980-386 Supplier Electron Microscopy Sciences Supplier No. 14022
Single Component B Durcupan, Water Soluble- a water soluble embedding medium for EM based on an aliphatic polyepoxide.