Learn More
MSC Durham Bin Shelving Unit, 36 Adjustable Compartments, Steel, Gray
Catalog No. 5000512977
Encompass
This parts bin features 36 adjustable compartments, providing versatile storage for various components. Constructed from durable steel with a gray powder coat gloss finish, it is designed for industrial storage applications. The unit has overall dimensions of 23-3/4 in (W) x 4-3/4 in (D) x 23-3/4 in (H) and a ship weight of 23 lb.
  • Constructed of prime cold-rolled steel.
  • Back panel allows wall mounting.
  • Fixed shelves are slotted to hold vertical dividers.
  • Vertical dividers are removable, allowing for multiple bin configurations.
  • Durable, gray, powder coat finish.
  • Compartment bottoms are angled to facilitate part removal.
  • Shelf fronts are 1-1/4 in high to retain parts and provide labeling areas.
Catalog No. 50-005-12977 Supplier MSC Supplier No. 89795900
This parts bin features 36 adjustable compartments, providing versatile storage for various components. Constructed from durable steel with a gray powder coat gloss finish, it is designed for industrial storage applications. The unit has overall dimensions of 23-3/4 in (W) x 4-3/4 in (D) x 23-3/4 in (H) and a ship weight of 23 lb.
  • Constructed of prime cold-rolled steel.
  • Back panel allows wall mounting.
  • Fixed shelves are slotted to hold vertical dividers.
  • Vertical dividers are removable, allowing for multiple bin configurations.
  • Durable, gray, powder coat finish.
  • Compartment bottoms are angled to facilitate part removal.
  • Shelf fronts are 1-1/4 in high to retain parts and provide labeling areas.