This 42-bin shelving unit efficiently organizes small parts for industrial, automotive, medical, and educational use. It features 42 gray steel bins with sloped shelves for easy content removal and powder-coated roll-formed sides for durability. Ships fully assembled.
- Equipped with 42 gray steel bins, each measuring 5-3/8 in W x 11-7/8 in D x 5-7/16 in H, providing extensive storage for various small items.
- Made from steel with a powder-coated finish for long-lasting performance.
- Sloped shelves and open design allow for easy content removal.
- Designed to hold labels for quick identification of parts.
- Roll-formed sides provide structural integrity and strength.
- Ships fully assembled, saving setup time.
- Overall dimensions of 33-3/4 in W x 12 in D x 42 in H make it suitable for various spaces.
- Country of origin: Mexico.