Learn More
MSC Durham Bin Shelving Unit with Openings, 72 Bin, 33-3/4 in Width, Steel
Catalog No. 5000536562
Encompass
This bin shelving unit offers organized storage with 72 steel bins. Designed with durability in mind, the unit features a powder-coated finish and ships fully assembled for immediate use in various settings.
  • Easy removal of contents
  • Holds labels and retains parts
  • Roll-formed sides
  • Sloped shelves
Catalog No. 50-005-36562 Supplier MSC Supplier No. 89828800
This bin shelving unit offers organized storage with 72 steel bins. Designed with durability in mind, the unit features a powder-coated finish and ships fully assembled for immediate use in various settings.
  • Easy removal of contents
  • Holds labels and retains parts
  • Roll-formed sides
  • Sloped shelves