This drawer cabinet is designed for high-density storage of small parts in industrial environments. Constructed from cold rolled steel, it features 96 drawers for organized and efficient storage. The unit arrives fully assembled, providing a robust solution for various organizational needs.
- Built for industrial use with interlocking and welded construction
- Drawers have an interlocking design for superior strength
- Full width handles and easy glide runners for smooth movement
- Positive stops to prevent spilling
- Each drawer is slotted on 1 inch centers to accept high-impact plastic dividers (includes 2 per drawer)
- Drawer capacity is 10 lbs.
- Total cabinet capacity not to exceed 500 lbs.
- Powder coat finish for durability
- Fully welded base raises unit 5-3/4 inches above the floor