Learn More
MSC Durham Heavy-Duty Service Utility Cart, 36 in Long, 24 in Wide, Steel, 1200 lb Capacity
Catalog No. 5000527053
Encompass
This heavy-duty service utility cart is constructed from durable steel and offers a robust 1200 lb load capacity, making it ideal for demanding applications. It features three spacious shelves for efficient storage and transport of various items.
  • Swivel casters provide excellent maneuverability.
  • Side brake ensures secure placement.
  • Lipped shelf profile helps contain contents.
  • Features 5 in wheel diameter.
  • Provides 12.56 in shelf clearance.
  • Comes in a gray finish.
  • Manufactured in Mexico.
Catalog No. 50-005-27053 Supplier MSC Supplier No. 43180587
This heavy-duty service utility cart is constructed from durable steel and offers a robust 1200 lb load capacity, making it ideal for demanding applications. It features three spacious shelves for efficient storage and transport of various items.
  • Swivel casters provide excellent maneuverability.
  • Side brake ensures secure placement.
  • Lipped shelf profile helps contain contents.
  • Features 5 in wheel diameter.
  • Provides 12.56 in shelf clearance.
  • Comes in a gray finish.
  • Manufactured in Mexico.