Learn More
MSC Durham Heavy-Duty Utility Cart, 1200 lb Capacity, Steel, 3 Shelves
Catalog No. 5000514665
Encompass
This heavy-duty service utility cart is constructed from steel and designed for robust use. It features a 1200 lb load capacity, 3 shelves, and swivel casters for easy maneuverability in various work environments.
  • Heavy-duty design
  • Swivel caster type
  • Steel material
  • 60 in length
  • 30 in width
  • 1200 lb load capacity
  • 5 in wheel diameter
  • 12.56 in shelf clearance
  • Lipped shelf profile
  • Gray color
  • 3 shelves
  • Country of origin: Mexico
Catalog No. 50-005-14665 Supplier MSC Supplier No. 43180678
This heavy-duty service utility cart is constructed from steel and designed for robust use. It features a 1200 lb load capacity, 3 shelves, and swivel casters for easy maneuverability in various work environments.
  • Heavy-duty design
  • Swivel caster type
  • Steel material
  • 60 in length
  • 30 in width
  • 1200 lb load capacity
  • 5 in wheel diameter
  • 12.56 in shelf clearance
  • Lipped shelf profile
  • Gray color
  • 3 shelves
  • Country of origin: Mexico