Learn More
MSC Durham Manufacturing 9Fx First Aid Cabinet, 3-Shelf, 20-Gauge, 1 ea
Catalog No. 5000796189
Encompass
This first aid cabinet is constructed from 20-gauge steel and features three fixed shelves and a convenient carrying handle. It is finished with a gloss white coating, offering durability and a clean aesthetic for various environments.
  • Three fixed shelves
  • Carrying handle
  • Constructed from 20-gauge steel
  • Gloss white finish
  • Overall dimensions of 15-3/16 x 5-9/16 x 16-7/16 in.
  • Shipping weight of 12 lbs
Catalog No. 50-007-96189 Supplier MSC Supplier No. 00657353
This first aid cabinet is constructed from 20-gauge steel and features three fixed shelves and a convenient carrying handle. It is finished with a gloss white coating, offering durability and a clean aesthetic for various environments.
  • Three fixed shelves
  • Carrying handle
  • Constructed from 20-gauge steel
  • Gloss white finish
  • Overall dimensions of 15-3/16 x 5-9/16 x 16-7/16 in.
  • Shipping weight of 12 lbs