Learn More
MSC Durham Manufacturing Flammable Storage Cabinet, 45 gal., 1 ea
Catalog No. 5000796144
Encompass
This flammable storage cabinet is designed for the safe and secure storage of flammable liquids. With a 45-gallon capacity and a double-wall steel construction, it provides robust protection in industrial and laboratory settings.
  • Two manual closing doors
  • Two adjustable shelves
  • Heavy duty steel material with powder coat gloss finish
  • Overall dimensions: 43-3/16 W x 18-3/16 D x 65-1/8 H in.
  • Shelf dimensions: 39-3/4 x 13-5/8 in.
  • Recessed, pad lockable handle with keyed lock
  • FM approved, meets NFPA 30 and OSHA standards
  • Safety yellow color
  • Country of origin: Mexico
Catalog No. 50-007-96144 Supplier MSC Supplier No. 03439304
This flammable storage cabinet is designed for the safe and secure storage of flammable liquids. With a 45-gallon capacity and a double-wall steel construction, it provides robust protection in industrial and laboratory settings.
  • Two manual closing doors
  • Two adjustable shelves
  • Heavy duty steel material with powder coat gloss finish
  • Overall dimensions: 43-3/16 W x 18-3/16 D x 65-1/8 H in.
  • Shelf dimensions: 39-3/4 x 13-5/8 in.
  • Recessed, pad lockable handle with keyed lock
  • FM approved, meets NFPA 30 and OSHA standards
  • Safety yellow color
  • Country of origin: Mexico