Learn More
Grainger DURHAM MFG Drawer Bin Cabinet: 33 3/4 in x 17 3/4 in x 21 in, 30 Drawers, Stackable, Steel, Gray
Catalog No. NC3937371
Encompass_Preferred
These small parts drawer cabinets can be stacked on top of other compatible small parts drawer cabinets to provide high-density storage. The cabinets have metal drawers and metal frames. They are more rigid than small parts cabinets with plastic drawers and plastic frames.
Catalog No. NC3937371 Supplier Grainger Supplier No. 3W400
These small parts drawer cabinets can be stacked on top of other compatible small parts drawer cabinets to provide high-density storage. The cabinets have metal drawers and metal frames. They are more rigid than small parts cabinets with plastic drawers and plastic frames.