Learn More
MSC Durham Mfg Storage Cabinet, 1 Shelf, Steel, 24-1/8 x 24-1/4 x 33-5/8 in
Catalog No. 5000528197
Encompass
This steel storage cabinet is a stationary workstation designed with one fixed shelf. It provides secure storage for various items in industrial and commercial environments.
  • Stationary workstation design.
  • Constructed from steel for durability.
  • Features one fixed shelf.
  • Gray powder coat, textured finish.
  • Keyed locking mechanism with 3 locking points.
  • Includes 4 keys.
  • Shelf load capacity of 100 lbs.
  • Cabinet is shipped fully assembled.
  • Overall dimensions: 24-1/8 in width x 24-1/4 in depth x 33-5/8 in height.
  • Shelf dimensions: 24 in width x 21-1/8 in depth.
Catalog No. 50-005-28197 Supplier MSC Supplier No. 33670688
This steel storage cabinet is a stationary workstation designed with one fixed shelf. It provides secure storage for various items in industrial and commercial environments.
  • Stationary workstation design.
  • Constructed from steel for durability.
  • Features one fixed shelf.
  • Gray powder coat, textured finish.
  • Keyed locking mechanism with 3 locking points.
  • Includes 4 keys.
  • Shelf load capacity of 100 lbs.
  • Cabinet is shipped fully assembled.
  • Overall dimensions: 24-1/8 in width x 24-1/4 in depth x 33-5/8 in height.
  • Shelf dimensions: 24 in width x 21-1/8 in depth.