Learn More
MSC Durham Mobile Table, Steel, 48 x 24 x 30 in, 1,200 lb Load Capacity
Catalog No. 5000514000
Encompass
The Durham Mobile Table is a durable steel work table designed for various industrial and laboratory applications. Featuring a robust construction and equipped with four wheels for easy mobility, this table provides a versatile solution for transporting and organizing materials.
  • Made from durable steel for long-lasting use.
  • Provides a 1,200 lb load capacity for heavy items.
  • Features a gray, textured finish.
  • Equipped with two shelves for organized storage.
  • Includes four wheels, two rigid and two swivel, made of polyurethane for easy mobility.
  • Casters are 5 x 1-1/4 in and made of hard rubber.
  • Measures 48 in long, 24 in wide, and 30 in high.
Catalog No. 50-005-14000 Supplier MSC Supplier No. 48603039
The Durham Mobile Table is a durable steel work table designed for various industrial and laboratory applications. Featuring a robust construction and equipped with four wheels for easy mobility, this table provides a versatile solution for transporting and organizing materials.
  • Made from durable steel for long-lasting use.
  • Provides a 1,200 lb load capacity for heavy items.
  • Features a gray, textured finish.
  • Equipped with two shelves for organized storage.
  • Includes four wheels, two rigid and two swivel, made of polyurethane for easy mobility.
  • Casters are 5 x 1-1/4 in and made of hard rubber.
  • Measures 48 in long, 24 in wide, and 30 in high.