Learn More
MSC Durham Small Parts Rack, For Large Compartment Boxes, Styrene Plastic, Gray
Catalog No. 5000533497
Encompass
This small parts rack is designed for organizing large compartment boxes, offering an efficient storage solution for various environments. It features a durable styrene plastic frame and a gray, powder-coated finish, ensuring longevity and resistance to wear. The rack is wall-mountable, providing versatile placement options to optimize workspace.
  • Designed for large compartment boxes
  • Gray color
  • Styrene plastic frame material
  • Wall mountable
  • Powder coated finish
Catalog No. 50-005-33497 Supplier MSC Supplier No. 00064964
This small parts rack is designed for organizing large compartment boxes, offering an efficient storage solution for various environments. It features a durable styrene plastic frame and a gray, powder-coated finish, ensuring longevity and resistance to wear. The rack is wall-mountable, providing versatile placement options to optimize workspace.
  • Designed for large compartment boxes
  • Gray color
  • Styrene plastic frame material
  • Wall mountable
  • Powder coated finish