Learn More
MSC Durham Small Parts Storage Bin, 60 Bin, 12 in D x 35 in W x 65 in H
Catalog No. 5000532521
Encompass
The Durham Small Parts Storage Bin offers an efficient and organized storage solution for various small components. Constructed with a durable frame and equipped with multiple bins, it is well-suited for industrial, laboratory, and workshop environments.
  • Sixty bins for systematic organization
  • Dimensions of 12 in deep x 35 in wide x 65 in high
  • Frame constructed from styrene plastic
  • Drawers made of steel
  • Powder coated finish for durability
  • Includes removable dividers for flexible storage configurations
  • Gray color
Catalog No. 50-005-32521 Supplier MSC Supplier No. 33732363
The Durham Small Parts Storage Bin offers an efficient and organized storage solution for various small components. Constructed with a durable frame and equipped with multiple bins, it is well-suited for industrial, laboratory, and workshop environments.
  • Sixty bins for systematic organization
  • Dimensions of 12 in deep x 35 in wide x 65 in high
  • Frame constructed from styrene plastic
  • Drawers made of steel
  • Powder coated finish for durability
  • Includes removable dividers for flexible storage configurations
  • Gray color