Learn More
MSC Durham Steel Storage Cabinet, 64 Plastic Drawers, Gray, 1 ea
Catalog No. 5000515723
Encompass
This steel storage cabinet features 64 plastic drawers, providing an efficient solution for organizing and storing small parts. Constructed from durable 20-gauge steel with a gray powder-coated finish, it is designed for reliable use in workshop and industrial environments.
  • Features 64 individual plastic drawers for categorized storage
  • Constructed from 20-gauge steel with a powder-coated finish for durability
  • Includes two easy to label dividers that can be placed horizontally or vertically
  • Overall dimensions are 25-7/8 in. W x 6-3/8 in. D x 21-7/16 in. H
Catalog No. 50-005-15723 Supplier MSC Supplier No. 89795983
This steel storage cabinet features 64 plastic drawers, providing an efficient solution for organizing and storing small parts. Constructed from durable 20-gauge steel with a gray powder-coated finish, it is designed for reliable use in workshop and industrial environments.
  • Features 64 individual plastic drawers for categorized storage
  • Constructed from 20-gauge steel with a powder-coated finish for durability
  • Includes two easy to label dividers that can be placed horizontally or vertically
  • Overall dimensions are 25-7/8 in. W x 6-3/8 in. D x 21-7/16 in. H