Learn More
MSC Norton Floor Edger Disc, Silicon Carbide, Heavy Paper Backing, for Wood
Catalog No. 502836102
Encompass
The Norton Durite Paper Floor Edger Disc features 100% silicon carbide abrasive with heavy E, F, and combination paper backings for a quality cut and strength on the job. This traditional abrasive grain provides value and consistent performance at a competitive price. It is ideal for soft to medium hardness of wood.
  • Heavy duty paper backing for quality cut and strength.
  • Value and consistent performance at a competitive price.
  • Reduces dish out of springwood on strip and plank floors.
  • Good choice for value and consistent wood edger jobs.
Catalog No. 50-283-6102 Supplier MSC Supplier No. 62025341
The Norton Durite Paper Floor Edger Disc features 100% silicon carbide abrasive with heavy E, F, and combination paper backings for a quality cut and strength on the job. This traditional abrasive grain provides value and consistent performance at a competitive price. It is ideal for soft to medium hardness of wood.
  • Heavy duty paper backing for quality cut and strength.
  • Value and consistent performance at a competitive price.
  • Reduces dish out of springwood on strip and plank floors.
  • Good choice for value and consistent wood edger jobs.