Learn More
MSC Norton Abrasives Floor Sanding Disc, 19 in Diameter, 2 in Arbor Hole, 60 Grit
Catalog No. 502837398
Encompass
The Norton Durite Paper Floor Sanding Disc is engineered for effective surface preparation on various flooring types. Featuring a 100% silicon carbide abrasive, these discs are designed for rapid material removal and consistent finishing. The heavy paper backing ensures durability and prolonged disc life, making them suitable for demanding sanding tasks on both wood and concrete surfaces. Ideal for blending scratches from edger and drum sanders, hard-plating, and final sanding applications.
  • Utilizes 100% silicon carbide abrasive for aggressive cutting.
  • Heavy paper backing provides enhanced durability and extended product life.
  • Suitable for preparing wood and concrete surfaces.
  • Effective for removing various residues and blending scratches.
  • Designed for efficient hard-plating and final sanding.
Catalog No. 50-283-7398 Supplier MSC Supplier No. 56490162
The Norton Durite Paper Floor Sanding Disc is engineered for effective surface preparation on various flooring types. Featuring a 100% silicon carbide abrasive, these discs are designed for rapid material removal and consistent finishing. The heavy paper backing ensures durability and prolonged disc life, making them suitable for demanding sanding tasks on both wood and concrete surfaces. Ideal for blending scratches from edger and drum sanders, hard-plating, and final sanding applications.
  • Utilizes 100% silicon carbide abrasive for aggressive cutting.
  • Heavy paper backing provides enhanced durability and extended product life.
  • Suitable for preparing wood and concrete surfaces.
  • Effective for removing various residues and blending scratches.
  • Designed for efficient hard-plating and final sanding.