Learn More
MSC Norton Durite Paper Double-Sided Disc, 16 in diameter, Silicon Carbide, Floor Sanding
Catalog No. 502839700
Encompass
Norton Durite Paper Double-Sided Discs are designed for floor sanding applications on hard woods or concrete, offering quality and consistency. These double-sided discs simplify hard plating by removing the need for mechanical fastening to the buffer's bottom. For optimal results, a screen driver pad should always be used to prevent slippage and achieve the flattest possible surface.
  • 100% silicon carbide abrasive on both sides for extended life, quality cutting, and blending.
  • Heavy E, F, and combination paper backing provides strength, long life, and tear resistance.
  • Reduces dish-out of springwood on strip and plank floors.
  • Offers great value for consistent surface preparation and residue removal.
Catalog No. 50-283-9700 Supplier MSC Supplier No. 75584094
Norton Durite Paper Double-Sided Discs are designed for floor sanding applications on hard woods or concrete, offering quality and consistency. These double-sided discs simplify hard plating by removing the need for mechanical fastening to the buffer's bottom. For optimal results, a screen driver pad should always be used to prevent slippage and achieve the flattest possible surface.
  • 100% silicon carbide abrasive on both sides for extended life, quality cutting, and blending.
  • Heavy E, F, and combination paper backing provides strength, long life, and tear resistance.
  • Reduces dish-out of springwood on strip and plank floors.
  • Offers great value for consistent surface preparation and residue removal.