Learn More
MSC Norton Drum Paper, Silicon Carbide, 8 x 20-3/16 in, 80 grit
Catalog No. 502836235
Encompass
The Norton Durite Silicon Carbide Drum Paper is designed for high-quality abrasive performance on various small job applications. It features a tough backing for durability and is pre-cut to fit manufacturer drum sander specifications, commonly used in the rental trade for easy attachment and effective first cuts.
  • 100% silicon carbide abrasive for quality cut on all phases of a small job.
  • Tough E and F-weight backing resists tearing and extends life.
  • Sheets are pre-cut to meet manufacturers drum sander specifications, popular in the rental trade.
  • Easily attaches to the sander drum, a good choice for the first cut.
Catalog No. 50-283-6235 Supplier MSC Supplier No. 61033627
The Norton Durite Silicon Carbide Drum Paper is designed for high-quality abrasive performance on various small job applications. It features a tough backing for durability and is pre-cut to fit manufacturer drum sander specifications, commonly used in the rental trade for easy attachment and effective first cuts.
  • 100% silicon carbide abrasive for quality cut on all phases of a small job.
  • Tough E and F-weight backing resists tearing and extends life.
  • Sheets are pre-cut to meet manufacturers drum sander specifications, popular in the rental trade.
  • Easily attaches to the sander drum, a good choice for the first cut.