Learn More
MSC Norton Abrasives Floor Edger Disc, Silicon Carbide, 7 in, 7/8 in Arbor Hole, 24 Grit
Catalog No. 502838863
Encompass
The Durite Paper Floor Edger Disc is engineered for high-performance floor sanding and grinding applications. It features a durable silicon carbide abrasive and a heavy paper backing, making it suitable for both soft and medium hardness wood floors.
  • Abrasive material is 100% silicon carbide
  • Features heavy paper backing for durability
  • Offers effective sanding for various wood types
  • Designed for floor sanding and grinding applications
  • Available in multiple diameters and arbor hole sizes
  • Comes in a comprehensive range of grit sizes
Catalog No. 50-283-8863 Supplier MSC Supplier No. 43561174
The Durite Paper Floor Edger Disc is engineered for high-performance floor sanding and grinding applications. It features a durable silicon carbide abrasive and a heavy paper backing, making it suitable for both soft and medium hardness wood floors.
  • Abrasive material is 100% silicon carbide
  • Features heavy paper backing for durability
  • Offers effective sanding for various wood types
  • Designed for floor sanding and grinding applications
  • Available in multiple diameters and arbor hole sizes
  • Comes in a comprehensive range of grit sizes