Learn More
MSC Norton Edger Sanding Disc, Silicon Carbide, 7-7/8 in, E/F-Weight Paper Backing
Catalog No. 502839988
Encompass
This coarse grit paper floor sanding disc is designed for consistent residue removal during surface preparation on wood or concrete. It features a 100 percent silicon carbide abrasive paired with E- and F-weight heavy paper backing, ensuring a long disc life. The design helps reduce dishing out of springwood on strip and plank floors and easily blends scratches from edger and drum sanders.
  • Designed for consistent residue removal during surface preparation
  • Made with 100 percent silicon carbide abrasive
  • Features E- and F-weight heavy paper backing for long disc life
  • Reduces dishing out of springwood on strip and plank floors
  • Easily blends scratches from edger and drum sanders
Catalog No. 50-283-9988 Supplier MSC Supplier No. 90212911
This coarse grit paper floor sanding disc is designed for consistent residue removal during surface preparation on wood or concrete. It features a 100 percent silicon carbide abrasive paired with E- and F-weight heavy paper backing, ensuring a long disc life. The design helps reduce dishing out of springwood on strip and plank floors and easily blends scratches from edger and drum sanders.
  • Designed for consistent residue removal during surface preparation
  • Made with 100 percent silicon carbide abrasive
  • Features E- and F-weight heavy paper backing for long disc life
  • Reduces dishing out of springwood on strip and plank floors
  • Easily blends scratches from edger and drum sanders