Learn More
MSC Norton Abrasives Edger Disc, Silicon Carbide, Heavy Paper Backing
Catalog No. 502840034
Encompass
The Norton Durite Paper Floor Edger Disc features a 100% silicon carbide abrasive with heavy E, F, and combination paper backings. This abrasive disc is designed for quality cutting and strength, making it ideal for sanding soft to medium hardness wood floors. It offers consistent performance and value for various wood edger applications.
  • 100% silicon carbide abrasive provides effective material removal.
  • Heavy duty paper backing ensures durability and strength during use.
  • Ideal for soft to medium hardness wood applications.
  • Reduces dish out of springwood on strip and plank floors.
  • Offers good value and consistent performance for wood edger jobs.
Catalog No. 50-284-0034 Supplier MSC Supplier No. 90306812
The Norton Durite Paper Floor Edger Disc features a 100% silicon carbide abrasive with heavy E, F, and combination paper backings. This abrasive disc is designed for quality cutting and strength, making it ideal for sanding soft to medium hardness wood floors. It offers consistent performance and value for various wood edger applications.
  • 100% silicon carbide abrasive provides effective material removal.
  • Heavy duty paper backing ensures durability and strength during use.
  • Ideal for soft to medium hardness wood applications.
  • Reduces dish out of springwood on strip and plank floors.
  • Offers good value and consistent performance for wood edger jobs.