Learn More
MSC Norton Paper Floor Edger Disc, 7 in Diameter, 7/8 in Arbor Hole, 24 Grit
Catalog No. 502840035
Encompass
The Norton Durite Paper Floor Edger Disc features a 100% silicon carbide abrasive with heavy E, F, and combination paper backings. It is designed for quality cut and strength. This traditional abrasive grain offers value and consistent performance at a competitive price. It is ideal for soft to medium hardness wood.
  • Heavy-duty paper backing for quality cut and strength.
  • Consistent performance at a competitive price.
  • Reduction of "dish out" of springwood on strip and plank floors.
  • Good choice for value and consistent wood edger jobs.
Catalog No. 50-284-0035 Supplier MSC Supplier No. 90212937
The Norton Durite Paper Floor Edger Disc features a 100% silicon carbide abrasive with heavy E, F, and combination paper backings. It is designed for quality cut and strength. This traditional abrasive grain offers value and consistent performance at a competitive price. It is ideal for soft to medium hardness wood.
  • Heavy-duty paper backing for quality cut and strength.
  • Consistent performance at a competitive price.
  • Reduction of "dish out" of springwood on strip and plank floors.
  • Good choice for value and consistent wood edger jobs.