Learn More
MSC Norton Paper Floor Sanding Disc, Silicon Carbide, Heavy Paper Backing
Catalog No. 502838637
Encompass
Norton Durite Paper Floor Sanding Discs are suitable for surface preparation on wood and concrete to remove various residues. These large-diameter discs offer aggressive cutting and can easily blend scratches from edger and drum sanders. They are used for hard-plating and final sanding to prevent dish-out of softer grains.
  • 100% silicon carbide abrasive for a quality cut, easily blending scratches from edger and drum sanders.
  • Heavy E, F, and combination paper backings provide strength and long disc life.
  • Reduces dish-out of springwood on strip and plank floors.
  • Offers good value and consistent performance.
Catalog No. 50-283-8637 Supplier MSC Supplier No. 73252850
Norton Durite Paper Floor Sanding Discs are suitable for surface preparation on wood and concrete to remove various residues. These large-diameter discs offer aggressive cutting and can easily blend scratches from edger and drum sanders. They are used for hard-plating and final sanding to prevent dish-out of softer grains.
  • 100% silicon carbide abrasive for a quality cut, easily blending scratches from edger and drum sanders.
  • Heavy E, F, and combination paper backings provide strength and long disc life.
  • Reduces dish-out of springwood on strip and plank floors.
  • Offers good value and consistent performance.