Learn More
MSC Norton Screen-Bak Durite Screen Floor Edger Disc, Silicon Carbide, 7 in, P60 Grit
Catalog No. 502838002
Encompass
When aggressive performance and good value are desired, these screen floor edger discs are a suitable choice. They feature an interwoven polyester knit backing and open mesh construction, providing strength and resistance to loading during floor sanding projects.
  • P-graded sharp silicon carbide offers excellent cutting and non-loading properties.
  • Waterproof; can be rinsed and reused for extended sanding life.
  • Open weave screen provides extreme resistance to loading when removing scale, rust, and corrosion.
  • More durable and tougher than paper or cloth abrasives.
Catalog No. 50-283-8002 Supplier MSC Supplier No. 75607176
When aggressive performance and good value are desired, these screen floor edger discs are a suitable choice. They feature an interwoven polyester knit backing and open mesh construction, providing strength and resistance to loading during floor sanding projects.
  • P-graded sharp silicon carbide offers excellent cutting and non-loading properties.
  • Waterproof; can be rinsed and reused for extended sanding life.
  • Open weave screen provides extreme resistance to loading when removing scale, rust, and corrosion.
  • More durable and tougher than paper or cloth abrasives.