Learn More
MSC Norton Screen Floor Edger Disc, Silicon Carbide, Screen-bak
Supplier: MSC 75607259
The Norton Screen-Bak Durite Screen Floor Edger Disc is designed for effective sanding. It features P-graded sharp silicon carbide abrasive, known for its excellent cutting and non-loading properties. The waterproof design allows for rinsing and reuse, extending the sanding life. Its open weave screen resists loading, making it ideal for removing scale, rust, and corrosion. This durable disc is tougher than traditional paper or cloth abrasives.
- P-graded sharp silicon carbide has excellent cutting and non-loading properties.
- Waterproof; rinse and reuse for longer sanding life.
- Open weave screen has extreme resistance to loading when removing scale, rust and corrosion.
- Tougher and more durable than paper or cloth abrasives.