Learn More
Medchemexpress LLC Durlobactam sodium | 1467157-21-6 | 99.6% | 299.24 g·mol⁻¹ | C8H10N3NaO6S | 1 MG
SDP
Catalog No. 5000226574
Encompass_Preferred
Durlobactam sodium is the monosodium salt of a broad-spectrum β-lactamase inhibitor used in preclinical biochemical and microbiological research to evaluate inhibition of class A, C, and D β-lactamases. Supplied as a high-purity solid for use in in vitro assays and combination studies with β-lactam antibiotics.
  • High purity suitable for research applications.
  • Broad-spectrum β-lactamase inhibition activity.
  • White to light yellow solid, easy to handle.
  • Stable when stored sealed and protected from moisture.
  • Available in small research pack sizes for assay development.
Catalog No. 50-002-26574 Supplier Medchemexpress LLC Supplier No. HY1179741MG
Durlobactam sodium is the monosodium salt of a broad-spectrum β-lactamase inhibitor used in preclinical biochemical and microbiological research to evaluate inhibition of class A, C, and D β-lactamases. Supplied as a high-purity solid for use in in vitro assays and combination studies with β-lactam antibiotics.
  • High purity suitable for research applications.
  • Broad-spectrum β-lactamase inhibition activity.
  • White to light yellow solid, easy to handle.
  • Stable when stored sealed and protected from moisture.
  • Available in small research pack sizes for assay development.